Learn More
Medchemexpress LLC Lascufloxacin | 848416-07-9 | 98.0% | 439.43 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY167451MG
Encompass_Preferred
Lascufloxacin is a potent and orally active fluoroquinolone antibacterial agent. It effectively inhibits infections caused by various pathogens, including quinolone-resistant strains. This product has the potential for treating various infectious diseases, notably lower respiratory tract infections.
  • Potent antibacterial activity against quinolone-resistant strains.
  • Effective against infections caused by various pathogens.
  • Potential for treating lower respiratory tract infections.
  • Shows antibacterial activities against *Moraxella catarrhalis* and *Haemophilus influenzae*.
  • Inhibits *E. coli* and *P. aeruginosa*.
  • Potent activity against macrolide-resistant *M. pneumoniae* isolates.
  • Exhibits significant bacterial killing in mouse models of infection.
Catalog No. 50-003-90927