Learn More
Sigma Aldrich Fine Chemicals Biosciences Laser Cryo-BabiesR green L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L37891PAK
Encompass
Label large numbers of 1.5AmL or 2AmL microcentrifuge tubes with the convenience and automation of word processing. Chemically inert labels adhere tightly and withstand autoclaving freezing (incl liquid nitrogen) most organic solvents and caustics. Sheets are specially top-coated to accept laser printing. Thin film and non-migrating adhesive mean that labeled tubes will not bind in centrifuge rotor.Withstand temperatures from -196C to 150C including liquid and vapor phase nitrogen boiling water baths (100C) autoclaving and moderate oven temperatures (150C)Chemically inert resist most organic solvents and caustic agentsWill adhere to most plastics glass and metals without cracking peeling or degradingHeat-resistant stay-flat sheets are specially top-coated to accept laser printing and will not jam in laser printersMade with a flexible polyolefinx2020 label material and acrylic adhesive. This unique combination expands and contracts together with temp
Catalog No. 50-290-1189