Learn More
Medchemexpress LLC Lasofoxifene tartrate | 190791-29-8 | 99.9% | C32H37NO8 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYA003810MM/1ML
Encompass_Preferred
Lasofoxifene (CP-336156) tartrate is an orally active and selective estrogen receptor modulator (SERM). It demonstrates anti-osteoporotic effects and inhibits primary tumor growth and metastases. It can be used for the research of breast cancer and postmenopausal osteoporosis.
  • Orally active and selective estrogen receptor modulator
  • Demonstrates anti-osteoporotic effects
  • Inhibits primary tumor growth and metastases
  • Useful for research of breast cancer
  • Useful for research of postmenopausal osteoporosis
Catalog No. 50-004-33164