Learn More
Cayman Chemical LasofoxIfentartrate 10mg
SDP
Supplier:  Cayman Chemical 1764610 MG
Encompass_Preferred
A third-generation SERM that selectively binds to human ERα (IC50 = 1.5 nM) and inhibits bone loss in ovariectomized rats; used in the treatment of postmenopausal osteoporosis and ER-positive breast cancer; also acts as an inverse agonist at the CB2 cannabinoid receptor
Catalog No. 50-296-6337