Learn More
TARGETMOL CHEMICALS INC LATANOPROST 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T252810MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Latanoprost (Xalatan) is a prostaglandin F2alpha analogue and a prostanoid selective FP receptor agonist with an ocular hypertensive effect. purity: 99%
Catalog No. 50-258-0761