Learn More
Medchemexpress LLC Latanoprost | 130209-82-4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB057710MM/1ML
Encompass_Preferred
Latanoprost is a prostaglandin F2α analogue used for glaucoma research. It passes through the cornea and is hydrolyzed by esterase to latanoprost acid, which is an F-prostaglandin (FP) receptor agonist. Latanoprost acid effectively reduces intraocular pressure (IOP) by increasing the outflow of aqueous humor through the uvea.
  • Prostaglandin F2α analogue
  • Used for glaucoma research
  • Passes through the cornea
  • Hydrolyzed by esterase to latanoprost acid
  • FP receptor agonist
  • Reduces intraocular pressure (IOP)
  • Increases outflow of aqueous humor through the uvea
Catalog No. 50-004-08672