Learn More
Medchemexpress LLC Latanoprost | 130209-82-4 | MFCD00864319 | >98% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0577
Encompass_Preferred
Latanoprost is a prostaglandin F2α analogue and an agonist for the FP prostanoid receptor. It is primarily used in glaucoma research to effectively reduce intraocular pressure (IOP) by increasing the outflow of aqueous humor through the uvea. It can effectively pass through the cornea and is hydrolyzed by esterase to latanoprost acid.
  • Prostaglandin F2α analogue
  • Agonist for the FP prostanoid receptor
  • Lowers intraocular pressure (IOP)
  • Inhibits VEGF-induced proliferation and migration in human umbilical vein endothelial cells (HUVECs)
  • Suppresses retinal neovascularization in vitro
  • Increases cell viability in HUVECs and human retinal microvascular endothelial cells (HRMECs)
  • Decreases levels of Akt, Erk1/2, NF-κB, and VEGF expression
  • Reduces blood flow in the optic nerve head, iris, and choroid
  • Inhibits the development of choroidal neovascularization in mice
Catalog No. 50-004-50821