Learn More
Selleck Chemical LLC Latanoprost S4709-25mg
SDP
Supplier:  Selleck Chemical LLC S470925MG
Encompass_Preferred
Latanoprost (Xalatan PhXA41) is a prostaglandin F2alpha analogue and a prostanoid selective FP receptor agonist with an ocular hypertensive effect Latanoprost increases uveoscleral outflow and thereby reduces intraocular pressure
Catalog No. 50-311-2056