Learn More
Medchemexpress LLC Latanoprost acid | 41639-83-2 | 95.1% | 390.51 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY113756A
Encompass_Preferred
Latanoprost acid is an analog of prostaglandin (PG) F2α and a selective prostanoid receptor (FP) agonist that activates the FP-PG receptor. It inhibits RANKL-induced osteoclastogenesis and function by inhibiting the ERK, AKT, JNK, and p38 cascade, following the c-fos/NFATc1 pathway. This medication is used to lower pressure inside the eyes and is for research use only.
  • Selective prostanoid receptor (FP) agonist
  • Activates the FP-PG receptor
  • Inhibits RANKL-induced osteoclastogenesis and function
  • Inhibits ERK, AKT, JNK, and p38 cascade
  • Soluble in DMSO at 100 mg/mL
Catalog No. 50-004-39179