Learn More
Medchemexpress LLC Latanoprost acid | 41639-83-2 | 95.1% | 390.51 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY113756A50MG
Encompass_Preferred
Latanoprost acid, an analog of prostaglandin (PG) F2α, is a selective prostanoid receptor (FP) agonist that specifically activates the FP-PG receptor. It inhibits RANKL-induced osteoclastogenesis and function by inhibiting ERK, AKT, JNK, and p38 cascade, followed by the c-fos/NFATc1 pathway. Latanoprost acid is a medication used to lower pressure inside the eyes.
  • Selective prostanoid receptor (FP) agonist
  • Specifically activates the FP-PG receptor
  • Inhibits RANKL-induced osteoclastogenesis and function
  • Inhibits ERK, AKT, JNK, and p38 cascade
  • Inhibits the c-fos/NFATc1 pathway
  • Lowers pressure inside the eyes
  • Reduces the protein expressions of c-fos and NFATc1 in vitro
  • Significantly inhibits mature osteoclast formation in vitro
  • Notably prevents LPS-induced bone destruction in vivo
Catalog No. 50-004-38472