Learn More
Medchemexpress LLC Latanoprost acid | 41639-83-2 | MFCD08056084 | 99.6% | 390.51 | C23H34O5 | 5 MG

Supplier: Medchemexpress LLC HY113756A5MG
Latanoprost acid is the free acid form of latanoprost, a prostaglandin F (FP) receptor agonist used as a research reagent and analytical reference standard. It is supplied in high purity for pharmacology, biochemical assays, and HPLC-MS/MS analysis, and is available as solid quantities and as a DMSO solution for assay preparation.
- High purity (99.62%).
- Molecular formula C23H34O5; molecular weight 390.51.
- Available in multiple pack sizes, including 5 mg and larger quantities.
- Soluble in DMSO; 10 mM solution offered for direct use in assays.
- Suitable as an FP receptor agonist for pharmacological and analytical studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.