Learn More
Medchemexpress LLC Latanoprost lactone diol | 145667-75-0 | MFCD05865275 | 99.9% | 304.38 | C18H24O4 | 50MG

Supplier: Medchemexpress LLC HY12594650MG
Latanoprost lactone diol is a chemical intermediate and analytical standard used in the synthesis and study of prostaglandin analogs such as latanoprost. Supplied for research use only, it is offered in solid and solution formats for medicinal chemistry and ophthalmic research applications.
- CAS number 145667-75-0.
- Purity 99.86% (reported by supplier).
- Molecular weight 304.38 g/mol.
- Available in multiple sizes, including 50 mg.
- Provided as solid and DMSO solution formats.
- Intended for research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.