Learn More
Medchemexpress LLC Latanoprost lactone diol | 145667-75-0 | MFCD05865275 | 99.9% | 304.38 g/mol | C18H24O4 | 100MG
SDP
Supplier:  Medchemexpress LLC HY125946100MG
Encompass_Preferred
Latanoprost lactone diol is a prostaglandin F-series intermediate supplied as a high-purity reference and synthetic building block for research. It is used in medicinal chemistry and analytical workflows, and is provided both as a solid and as ready-made DMSO solutions for convenience.
  • High purity: 99.9% (manufacturer reported)
  • Molecular formula C18H24O4 and molecular weight 304.38 g/mol
  • Available as solid and 10 mM solutions in DMSO
  • Multiple sizes including a 100 mg option for small-scale work
  • Intended for research and synthesis use only, not for human use
Catalog No. 50-002-35342