Learn More
Medchemexpress LLC Latanoprost lactone diol | 145667-75-0 | MFCD05865275 | 99.9% | 304.38 g/mol | C18H24O4 | 10MM 1ML

Supplier: Medchemexpress LLC HY12594610MM 1ML
Latanoprost lactone diol is a synthetic intermediate used in the preparation of latanoprost, a prostaglandin F2α analogue and FP receptor agonist that lowers intraocular pressure. The compound is provided for research purposes in ready-to-use solution or solid form and is characterized for purity and molecular identity.
- Intermediate in prostaglandin synthesis
- Supplied as a 10 mM solution in DMSO, 1 mL
- Also available as a solid with solvent for reconstitution
- High purity (99.86% reported)
- Molecular weight 304.38 g/mol
- Intended for research use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.