Learn More
Medchemexpress LLC Latanoprost lactone diol | 145667-75-0 | MFCD05865275 | 99.9% | 304.38 g·mol⁻¹ | C18H24O4 | 25MG

Supplier: Medchemexpress LLC HY12594625MG
Latanoprost lactone diol is a high-purity chemical intermediate used in the synthesis of latanoprost and for related research applications. The material is provided as solid quantities and as a DMSO solution, and is intended for synthetic workflows, analytical method development, and laboratory research requiring an intermediate of the prostaglandin F2α family.
- High purity: 99.86% reported
- Chemical formula: C18H24O4
- Molecular weight: 304.38 g·mol⁻¹
- Available sizes include 5 mg, 10 mg, 25 mg, 50 mg, and 100 mg
- Also available as a 10 mM solution in DMSO (1 mL)
- Intended use as a synthesis intermediate and research reagent
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.