Learn More
Medchemexpress LLC Latanoprost lactone diol | 145667-75-0 | MFCD05865275 | 99.9% | 304.38 g/mol | C18H24O4 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1259465MG
Encompass_Preferred
Latanoprost lactone diol is a research-grade intermediate and analytical standard (CAS 145667-75-0) used in the synthesis and impurity profiling of latanoprost and related prostaglandin analogs. Supplied as a high-purity crystalline material, it is intended for laboratory research, method development, and reference standard use.
  • High purity (≈99.9%) suitable for analytical applications.
  • Used as an intermediate in latanoprost synthesis.
  • Suitable for method development and validation.
  • Available in small pack sizes for analytical work.
  • Supplied as a crystalline solid for ease of handling.
  • Molecular formula C18H24O4; molecular weight 304.38 g/mol.
Catalog No. 50-002-35343