Learn More
Medchemexpress LLC HY-19518 5mg Medchemexpress, Latanoprostene bunod CAS:860005-21-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY195185MG
Encompass_Preferred
Medchemexpress, HY-19518 5mg Latanoprostene bunod CAS:860005-21-6 Latanoprostene bunod (LBN), a nitric oxide (NO)-donating prostaglandin F2a analog, is a topical ophthalmic therapeutic for the reduction of intraocular pressure (IOP) in patients with open-angle glaucoma or ocular hypertension (OHT) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2836