Learn More
Medchemexpress LLC Latent TGF beta 1/TG 100ug | 100ug
SDP
Supplier:  Medchemexpress LLC HYP78360100UG
Encompass_Preferred
Transforming growth factor beta-1 proprotein is the precursor of the Latency-associated peptide (LAP) and Transforming growth factor beta-1 (TGF-beta-1) chains TGF beta 1/TGFB1 Protein Mouse (HEK293 His) is a recombinant protein (L30-S390) produced by HEK293 cells with His tag[1][2]
Catalog No. 50-001-93915