Learn More
MOLECULAR DEPOT LLC Latex Blocking Buffer - 1 L
Supplier:  MOLECULAR DEPOT LLC B2010005L
Encompass_Preferred
Latex Microparticles Blocking Buffer (1 L) is an optimized versatile buffer for the conjugation of antibodies proteins peptides and small molecules to polystyrene (Latex) microparticles and nanoparticles This buffer is used to block of particles that have been activated with the standard EDC/NHS method Blocking particles after conjugation is essential for the stability and the specificity of the conjugated microparticles For in vitro research use only
Catalog No. 50-000-13540