Learn More
Cayman Chemical Lathosterol 10mg
SDP
Supplier:  Cayman Chemical 900310210 MG
Encompass_Preferred
An intermediate in the biosynthesis of cholesterol; serum levels of lathosterol have been used as biomarkers of excess cholesterol production; accumulates in lathosterolosis
Catalog No. 50-289-6017