Learn More
Cayman Chemical Latnoprost ethyl amIde-d4 25ug
SDP
Supplier:  Cayman Chemical 31682225 UG
Encompass_Preferred
An internal standard for the quantification of latanoprost ethyl amide by GC- or LC-MS.
Catalog No. 50-298-8059