Learn More
Cayman Chemical Latnoprost Lacton DIol 10mg
SDP
Supplier:  Cayman Chemical 7003910 MG
Encompass_Preferred
Latanoprost is an F-series prostaglandin analog which has been approved for use as an ocular hypotensive drug. Latanoprost lactone diol is an intermediate in the synthesis of latanoprost. Latanoprost lactone diol can be converted to the free acid of latanoprost by reduction with DIBAL followed by Wittig reaction with commercially available reagents.
Catalog No. 50-289-4268