Learn More
Cayman Chemical Latnoprosten Bunod 5mg
SDP
Supplier:  Cayman Chemical 258305 MG
Encompass_Preferred
A nitric oxide-donating FP receptor agonist; induces cGMP accumulation in PC12 and HEK293 cells (EC50s = 1.6 and 9.2 μM, respectively); topical administration reduces IOP in canine and rabbit models of glaucoma at 0.036%; decreases IOP in a cynomolgus monkey model of laser-induced ocular hypertension
Catalog No. 50-293-3422