Learn More
Cayman Chemical Latnoprostfree acId-d4 100ug
SDP
Supplier:  Cayman Chemical 316811100 UG
Encompass_Preferred
An internal standard for the quantification of latanoprost (free acid) by GC- or LC-MS
Catalog No. 50-298-8061