Learn More
FUJIFILM BIOSCIENCES INC Latrunculin A
Supplier:  FUJIFILM BIOSCIENCES INC 12504363
Encompass_Preferred
Latrunculin A is an actin polymerization inhibitor It binds to actin in a 1 1 ratio and strongly inhibits its polymerization and has been reported as an inhibitor of fertilization division and early development in sea urchins and mice For research use only RUO
Catalog No. 50-004-50284