Learn More
Medchemexpress LLC HY-138489 10mg , Lats-IN-1 CAS:1424635-83-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13848910MG
Encompass_Preferred
HY-138489 10mg Lats-IN-1 CAS: 1424635-83-5 Lats-IN-1 is a potent and ATP-competitive inhibitor of Lats1 and Lats2 kinases. Lats-IN-1 promotes Yap-dependent proliferation in postmitotic mammalian tissues. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9251