Learn More
Electron Microscopy Sciences Lattice Scriber for Semiconductor Wafers
SDP
Supplier:  Electron Microscopy Sciences 7645
Encompass_Preferred
This is the best scriber you'll ever find for scribing semiconductor wafers. Lattice Scriber is the only scriber that has a 8-point truncated diamond tip that can be used for both toe and heel scribing held by a robust 4" long pin vise handle.
Catalog No. 50-190-2665