Learn More
Lauda Brinkmann Lp LAUDA Universa U 8 M
Supplier:  Lauda Brinkmann Lp L003861
Encompass_Preferred
LAUDA Universa U 8 MHeating thermostat 100-125 V 50/60 HzTechnical FeaturesWorking temperature range 70 300 CWorking temperature range with external cooling 20 300 COperating temperature range -30 300 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 1 4 2 0 kWCurrent max 16 ASize of bath (W x D x H) 150 x 150 x 200 mm Bath opening (WxT) 150 x 150 mmBath volume min / max 5 8 / 8 5 LPump Pressure max 0 7 barPump suction max 0 4 barPump flow rate max (pressure) 25 L/minPump Flow rate max (suction) 23 L/minIn / Outlet connection thread (outside) M16 x 1In / Outlet olives 13 mmOverall dimensions (WxDxH) 230 x 400 x 497 mmWeight 18 kgPower supply 100-125 V 50/60 HzPower plug Power cord with plug (NEMA 5-20P)
Catalog No. 50-005-73948