Learn More
Medchemexpress LLC HY-D0080 10mg Medchemexpress, Laurdan CAS:74515-25-6 Purity:>98%
Supplier: Medchemexpress LLC HYD008010MG
Medchemexpress, HY-D0080 10mg Laurdan CAS:74515-25-6 Laurdan is a membrane-permeable fluorescent probe that displays spectral sensitivity to the phospholipid phase of the cell membrane to which it is bound. Quantitation of generalized polarization (GP) of Laurdan can be used to identify phospholipid phase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.