Learn More
Andwin Scientific LAURETH 12 50GR BOTTLE
SDP
Supplier:  Andwin Scientific 28144005
Encompass_Preferred
The Laureth 12 is ideal for use in elution solution. It is made to be utilized with Envirochek and Envirochek HV Sampling Capsules 28143-552 28145-715 -722 and -720. This product is packaged and sold individually in a 50 gram bottle.
Catalog No. NC9253856