Learn More
Cayman Chemical LaurIc AcId-13C 500mg
SDP
Supplier:  Cayman Chemical 29462500 MG
Encompass_Preferred
An internal standard for the quantification of lauric acid by GC- or LC-MS
Catalog No. 50-298-9202