Learn More
Cayman Chemical LaurIc AcId-d23 100mg
SDP
Supplier:  Cayman Chemical 28080100 MG
Encompass_Preferred
An internal standard for the quantification of lauric acid by GC- or LC-MS
Catalog No. 50-293-8899