Learn More
Cayman Chemical Lauric Acid Fatty Acids and De
SDP
Supplier:  Cayman Chemical 1000662625 G
Encompass_Preferred
A medium-chain fatty acid; has been found at high levels in coconut oil; induces the activation of NF-κB and the expression of COX-2, iNOS, and IL-1α in RAW 264.7 cells at 25 µM
Catalog No. 50-288-9850