Learn More
Medchemexpress LLC Laurolitsine hydrochloride | 100 MG

Supplier: Medchemexpress LLC HYN2352A100MG
Laurolitsine hydrochloride, also known as (+)-Norboldine hydrochloride, is an alkaloid isolated from the leaves of Peumus boldus Molina. It shows weak anti-inflammatory activity, particularly against nitric oxide (NO) production in RAW 267.4 and BV-2 cells, and also affects platelet aggregation.
- Isolated from the leaves of Peumus boldus Molina
- Exhibits weak anti-inflammatory activity
- Inhibits NO production in RAW 267.4 and BV-2 cells
- Inhibits aggregation of rabbit platelets induced by arachidonic acid and collagen
- Slightly inhibits aggregation induced by ADP
- Purity of 99.97%
- Molecular formula: C18H20ClNO4
- Molecular weight: 349.81
- Appears as a white to pink solid
- Soluble in DMSO at 100 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.