Learn More
Medchemexpress LLC Lauroyl lysine 50g | 52315-75-0 | 328.49 g/mol | C18H36N2O3 | 50 G

Supplier: Medchemexpress LLC HY11719550G
Lauroyl lysine is an amino acid derivative supplied as a reagent for laboratory research and formulation work. It is provided as a high-purity powder suitable for chemical synthesis and product development.
- Amino acid derivative, N6-lauroyl-L-lysine.
- High purity: 98.0%.
- Molecular weight: 328.49 g/mol.
- Chemical formula: C18H36N2O3.
- Available in multiple pack sizes, including 50 g.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.