Learn More
Medchemexpress LLC Lauroyl polyoxyl-32 glycerides | 121548-04-7 | MFCD30182354 | 100g
SDP
Supplier:  Medchemexpress LLC HYY1892100G
Encompass_Preferred
Gelucire 44/14 is a potential and safe absorption enhancer for improving the absorption of poorly absorbable agents including insulin and calcitonin by pulmonary delivery
Catalog No. 50-002-66405