Learn More
Cayman Chemical LauroylDLCarnItInchlorId 500mg
SDP
Supplier:  Cayman Chemical 16006500 MG
Encompass_Preferred
A zwitterionic, long-chain acylcarnitine used to improve in vivo absorption of certain hydrophilic compounds, especially through mucosal membranes
Catalog No. 50-296-9826