Learn More
Cayman Chemical LauroylLcarnItIn12 12 12 10mg
SDP
Supplier:  Cayman Chemical 2676910 MG
Encompass_Preferred
An internal standard for the quantification of lauroyl-L-carnitine by GC- or LC-MS
Catalog No. 50-293-2936