Learn More
Cayman Chemical LauroylLcarnItInchlorIde 100mg
SDP
Supplier:  Cayman Chemical 26550100 MG
Encompass_Preferred
An acylcarnitine and a quaternary ammonium-containing cationic surfactant; has been used to permeabilize porcine enterocytes for delivery of the polar fluorescent probe lucifer yellow
Catalog No. 50-293-6919