Learn More
Medchemexpress LLC Lauryl benzalkonium-d7 chloride | 1998128-83-8 | 347.03 | C21H31D7ClN | 10 MG

Supplier: Medchemexpress LLC HYB1549S10MG
Lauryl benzalkonium-d7 chloride is the deuterium-labeled analogue of lauryl benzalkonium chloride, a quaternary ammonium cationic surfactant used as an internal standard or tracer in analytical and biochemical research. The labeled compound facilitates mass spectrometric differentiation and quantitative studies and is supplied for research use only.
- Deuterium-labeled cationic surfactant.
- Molecular formula C21H31D7ClN.
- Molecular weight 347.03 g/mol.
- Supplied as a 10 mg vial.
- For research use only; not for human or clinical use.
- Useful as an internal standard for mass spectrometry-based quantitation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.