Learn More
Selleck Chemical LLC Lauryl gallate
SDP
Supplier:  Selleck Chemical LLC S622925MG
Encompass_Preferred
Lauryl gallate (Dodecyl gallate) is the ester of dodecanol and gallic acid that exhibits both potent chain-breaking and preventive antioxidant activity
Catalog No. 50-000-03280