Learn More
Cayman Chemical Lavendustin A Inhibitors
SDP
Supplier:  Cayman Chemical 100102681 MG
Encompass_Preferred
A selective inhibitor of EGF receptor-associated tyrosine kinase (IC50 = 11 nM); used to differentiate rat mesenchymal stem cells, to inhibit NMDA-stimulated cGMP production, and to inhibit VEGF-induced angiogenesis
Catalog No. 50-288-8580