Learn More
Medchemexpress LLC Lavendustin c | 125697-93-0 | MFCD00153955 | 98.3% | 275.26 g/mol | C14H13NO5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW01385710MG
Encompass_Preferred
Lavendustin C is a research-grade small-molecule kinase inhibitor used in biochemical and cellular studies. It is reported to potently inhibit Ca2+/calmodulin-dependent kinase II and to inhibit certain receptor-associated tyrosine kinases. The compound is provided as a solid for laboratory use, with molecular formula C14H13NO5 and molecular weight 275.26 g/mol.
  • Potent CaMK II inhibitor (reported IC50 ~0.2 μM).
  • Also inhibits EGFR-associated tyrosine kinase activity.
  • High purity suitable for research applications.
  • Supplied as a solid, soluble in DMSO for assay preparation.
  • Useful for biochemical assays and cellular signaling studies.
Catalog No. 50-003-03310