Learn More
Medchemexpress LLC Lawsone methyl ether | 2348-82-5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN711650MG
Encompass_Preferred
Lawsone methyl ether (2-Methoxy-1,4-naphthoquinone) is isolated from Impatiens balsamina L. and Swertia calycina. It shows potent antifungal and antibacterial activities and is for research use only.
  • Isolated from Impatiens balsamina L. and Swertia calycina.
  • Exhibits potent antifungal activities.
  • Exhibits potent antibacterial activities.
  • Shown cytotoxicity against various human cell lines.
  • Provided as a solid form.
  • Has a light yellow to yellow appearance.
  • Purity is 99.16%.
Catalog No. 50-004-17519