Learn More
Medchemexpress LLC Lazabemide (Ro 19-6327) | 103878-84-8 | 99.95% | C8H10ClN3O | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14201100MG
Encompass_Preferred
Lazabemide (Ro 19-6327) is a selective, reversible inhibitor of monoamine oxidase B (MAO-B). It shows less activity for MAO-A and can inhibit monoamine uptake at higher concentrations. This compound is suitable for research concerning Parkinson's and Alzheimer's disease.
  • Selective, reversible inhibitor of MAO-B
  • Less active for MAO-A
  • Inhibits monoamine uptake at high concentrations
  • Useful in Parkinson's disease research
  • Useful in Alzheimer's disease research
Catalog No. 50-003-53606