Learn More
Selleck Chemical LLC Lazertinib S8724-25MG
SDP
Supplier:  Selleck Chemical LLC S872425MG
Encompass_Preferred
Lazertinib (YH25448 GNS-1480) is a potent highly mutant-selective and irreversible EGFR-TKI with IC50 values of 1 7 nM 2 nM 5 nM 20 6 nM and 76 nM for Del19/T790M L858R/T790M Del19 L85R and Wild type EGFR respectively showing much higher IC50 values aganist ErbB2 and ErbB4
Catalog No. 50-000-24239