Learn More
Selleck Chemical LLC LB-100
SDP
Supplier:  Selleck Chemical LLC S753725MG
Encompass_Preferred
LB-100 is a water soluble protein phosphatase 2A (PP2A) inhibitor with IC50s of 0 85 M and 3 87 M in BxPc-3 and Panc-1 cells
Catalog No. 50-000-00069