Learn More
AG Scientific Inc LB Agar Low Salt Powder 500 G
Supplier: AG Scientific Inc A2799500GM
For propagation and maintenance of E. Coli. Low Salt Formula. Molecular Biology Grade. Protocol: Dissolve 35 g in 1 L distilled water. Adjust the pH to 7.2 Sterilize by autoclaving for 15 minutes