Learn More
Teknova LB Agar Plates with 1% Glucose and Chloramphenicol-34. 245mm, 4 Plates per Sleeve, Sterile. 4 Pack.
SDP
Supplier:  Teknova L68074PK
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.0% Cerelose Anhydrous Dextrose (Glucose), 1.5% Agar, 34g/mL Chloramphenicol
Catalog No. 50-238-7928