Learn More
Teknova LB Agar Plates with Carbenicillin-75 and Chloramphenicol-30. 100mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Supplier:  Teknova L12208PK
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 75g/mL Carbenicillin Disodium Salt, 30g/mL Chloramphenicol
Catalog No. 50-238-7842